AC5388701897 Accela ChemBio Inc.

5-Bromo-2-chloro-1,3-dinitrobenzene

  • MDL number MFCD22682798
  • Cas Number : 51796-82-8
  • Molecular Weight : 281.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022400&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY022400.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart