AC5388701899 Accela ChemBio Inc.

6,7-Dihydropyrido[2,3-d]pyridazine-5,8-dione

  • MDL number MFCD06251613
  • Cas Number : 4430-77-7
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022431&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022431.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart