AC5388701902 Accela ChemBio Inc.

N-Methyl-DL-leucine

  • MDL number MFCD00037758
  • Cas Number : 2566-33-8
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022445&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022445.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart