AC5388701908 Accela ChemBio Inc.

5-(Ethoxymethyl)-8-hydroxyquinoline Hydrochloride

  • MDL number MFCD22682794
  • Cas Number : 1418117-78-8
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022468&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022468.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart