AC5388701910 Accela ChemBio Inc.

1-(4-Bromo-2-methylphenyl)piperazine

  • MDL number MFCD03444506
  • Cas Number : 501903-60-2
  • Molecular Weight : 255.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022473&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022473.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart