AC5388701913 Accela ChemBio Inc.

3-Bromo-4-hydroxypyridine

  • MDL number MFCD01646107
  • Cas Number : 36953-41-0
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022509&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY022509.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart