AC5388701922 Accela ChemBio Inc.

(S,E)-2-Methyl-N-(2,2,2-trifluoroethylidene)propane-2-sulfinamide

  • MDL number MFCD20487958
  • Cas Number : 929642-48-8
  • Molecular Weight : 201.2099096

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022547&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022547.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart