AC5388701925 Accela ChemBio Inc.

4-Bromo-3-(bromomethyl)benzonitrile

  • MDL number MFCD12755973
  • Cas Number : 190197-86-5
  • Molecular Weight : 274.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022576&attname=GoodsCode

Safety Information

signalword Warning
MSDS http://pdf.accelachem.com/cn/msds/SY022576.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart