AC5388701926 Accela ChemBio Inc.

Ethyl 5-Aminothieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD22683327
  • Cas Number : 59488-80-1
  • Molecular Weight : 223.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022583&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022583.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart