AC5388701940 Accela ChemBio Inc.

2-[(4,4-Diethoxy-2-butyn-1-yl)oxy]tetrahydropyran

  • MDL number MFCD23381110
  • Cas Number : 99805-29-5
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022667&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022667.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart