AC5388701941 Accela ChemBio Inc.

2-(Aminomethyl)-5-nitrothiophene Hydrochloride

  • MDL number MFCD23381111
  • Cas Number : 1418117-90-4
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022669&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022669.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart