AC5388701942 Accela ChemBio Inc.

Bis(4-tetrahydropyranyl)amine

  • MDL number MFCD11841081
  • Cas Number : 1080028-76-7
  • Molecular Weight : 185.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022675&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P330+P331-P280-P305+P351+P338-P310-P301+P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY022675.pdf
hstatements H302-H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart