AC5388701956 Accela ChemBio Inc.

3-(2-Pyridyl)-1-propanol Hydrochloride

  • MDL number MFCD23381121
  • Cas Number : 52225-87-3
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023125&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023125.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart