AC5388701958 Accela ChemBio Inc.

N-[3-(Dimethylamino)-2-{[(dimethylamino)methylene]amino}prop-2-en-1-ylidene]-N-methylmethanaminium Hydrogen Dihexafluorophosphate

  • MDL number MFCD27955940
  • Cas Number : 295316-07-3
  • Molecular Weight : 488.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023204&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023204.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart