AC5388701962 Accela ChemBio Inc.

1-Benzylpyrazole-3-carbonitrile

  • MDL number MFCD23381125
  • Cas Number : 1427475-28-2
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023345&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023345.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart