AC5388701966 Accela ChemBio Inc.

3-(Aminomethyl)-1-benzylpyrazole Dihydrochloride

  • MDL number MFCD23703523
  • Cas Number : 1427475-21-5
  • Molecular Weight : 260.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023605&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023605.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart