AC5388701968 Accela ChemBio Inc.

3-Amino-5-(trifluoromethyl)pyrazole Hydrochloride

  • MDL number MFCD18633449
  • Cas Number : 1418117-74-4
  • Molecular Weight : 187.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022846&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022846.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart