AC5388701976 Accela ChemBio Inc.

1-(3-Pyridyl)-1-propylamine Dihydrochloride

  • MDL number MFCD09028142
  • Cas Number : 1228879-41-1
  • Molecular Weight : 209.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023048&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023048.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart