AC5388701982 Accela ChemBio Inc.

N-Cbz-pyrrolidine-3-carboxylic Acid

  • MDL number MFCD02179036
  • Cas Number : 188527-21-1
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023067&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY023067.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart