AC5388701988 Accela ChemBio Inc.

Ethyl 2-(4-Thiazolyl)acetate

  • MDL number MFCD03011394
  • Cas Number : 120155-43-3
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023096&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023096.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart