AC5388701989 Accela ChemBio Inc.

3,4-Dibromofuran

  • MDL number MFCD01318995
  • Cas Number : 32460-02-9
  • Molecular Weight : 225.86608

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023104&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P280-P403+P235-P501
hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY023104.pdf
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart