AC5388701997 Accela ChemBio Inc.

5-Methylthiophene-2,3-dicarboxylic Acid

  • MDL number MFCD14705596
  • Cas Number : 46029-22-5
  • Molecular Weight : 186.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023152&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023152.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart