AC5388702004 Accela ChemBio Inc.

2,5-Diazabicyclo[2.2.2]octane Dihydrochloride

  • MDL number MFCD26954797
  • Cas Number : 1192-92-3
  • Molecular Weight : 185.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023209&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023209.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart