AC5388702005 Accela ChemBio Inc.

3-Chloro-5-fluorobenzylamine

  • MDL number MFCD06213787
  • Cas Number : 90390-33-3
  • Molecular Weight : 159.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023211&attname=GoodsCode

Safety Information

signalword Danger
pcodes P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501A
hcodes GHS05
MSDS http://pdf.accelachem.com/cn/msds/SY023211.pdf
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart