AC5388702009 Accela ChemBio Inc.

4-Amino-1,3-dihydroisobenzofuran

  • MDL number MFCD16659158
  • Cas Number : 98475-10-6
  • Molecular Weight : 135.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023225&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023225.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart