AC5388702017 Accela ChemBio Inc.

4-Bromo-2-methoxybenzotrifluoride

  • MDL number MFCD13194486
  • Cas Number : 944805-63-4
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023303&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023303.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart