AC5388702031 Accela ChemBio Inc.

N-Boc-4-(methylsulfonyl)benzylamine

  • MDL number MFCD21106154
  • Cas Number : 694480-96-1
  • Molecular Weight : 285.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023506&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023506.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart