AC5394386031 Accela ChemBio Inc.

Methyl 2-Amino-2-(4-pyridyl)acetate Dihydrochloride

  • MDL number MFCD23703524
  • Cas Number : 1427475-14-6
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023726&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart