AC5394386034 Accela ChemBio Inc.

1,2,4-Tris(methylsulfonyl)benzene

  • MDL number MFCD24335247
  • Cas Number : 1434128-54-7
  • Molecular Weight : 312.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023802&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023802.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart