AC5394386037 Accela ChemBio Inc.

Methyl N-Boc-3-methylmorpholine-3-carboxylate

  • MDL number MFCD25292791
  • Cas Number : 1447240-83-6
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024195&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024195.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart