AC5394386042 Accela ChemBio Inc.

3-Aminoisonicotinamide Monohydrate

  • MDL number MFCD24842551
  • Cas Number : 1434128-46-7
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Pale yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024370&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024370.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart