AC5394386043 Accela ChemBio Inc.

Di(1-piperazinyl)methanone Dihydrochloride

  • MDL number MFCD24842552
  • Cas Number : 208711-30-2
  • Molecular Weight : 271.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024462&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024462.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart