AC5394386044 Accela ChemBio Inc.

Ethyl 5-Ethoxy-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

  • MDL number MFCD25292792
  • Cas Number : 23699-62-9
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024614&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024614.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart