AC5394386045 Accela ChemBio Inc.

1-Benzyl-3-ethoxypyrrolidine

  • MDL number MFCD24673464
  • Cas Number : 46441-02-5
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024622&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY024622.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart