AC5394386125 Accela ChemBio Inc.

8-Chloro-2-(methylthio)pyrimido[5,4-d]pyrimidine

  • MDL number MFCD00227587
  • Cas Number : 176637-10-8
  • Molecular Weight : 212.6594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011322&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011322.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart