AC5394386131 Accela ChemBio Inc.

2-Acetamidobenzyl Alcohol

  • MDL number MFCD14648693
  • Cas Number : 20939-77-9
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011672&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011672.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart