AC5394386133 Accela ChemBio Inc.

Samarium(III) Carbonate Hydrate

  • MDL number MFCD00150444
  • Cas Number : 38245-37-3
  • Molecular Weight : 498.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011799&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY011799.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart