AC5394386135 Accela ChemBio Inc.

2-(Hydroxymethyl)homomorpholine

  • MDL number MFCD18071030
  • Cas Number : 1207254-23-6
  • Molecular Weight : 131.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011928&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011928.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart