AC5394386138 Accela ChemBio Inc.

1-Benzyl-5-(hydroxymethyl)piperidin-2-one

  • MDL number MFCD18651683
  • Cas Number : 744212-68-8
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012353&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012353.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart