AC5394386142 Accela ChemBio Inc.

5-(2-Fluoro-5-nitrophenyl)-2H-tetrazole

  • MDL number MFCD18375288
  • Cas Number : 1171938-74-1
  • Molecular Weight : 209.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012444&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012444.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart