AC5394386166 Accela ChemBio Inc.

1-(Ethoxycarbonyl)-3-cyclopentene-1-carboxylic Acid

  • MDL number MFCD04115568
  • Cas Number : 76910-08-2
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014420&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014420.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart