AC5394386174 Accela ChemBio Inc.

5-[3-(Trifluoromethyl)phenyl]thiophene-2-carbaldehyde

  • MDL number MFCD06410192
  • Cas Number : 886508-92-5
  • Molecular Weight : 256.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015214&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015214.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart