AC5394386177 Accela ChemBio Inc.

6-(2,3-Dimethylphenyl)-3-pyridinecarbaldehyde

  • MDL number MFCD04115468
  • Cas Number : 924817-85-6
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015519&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015519.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart