AC5394386179 Accela ChemBio Inc.

5-tert-Butyl-2-hydroxybenzoic Acid

  • MDL number MFCD00156971
  • Cas Number : 16094-31-8
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015700&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015700.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart