AC5394386184 Accela ChemBio Inc.

Ethyl 5-Chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

  • MDL number MFCD11109466
  • Cas Number : 800401-62-1
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016185&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016185.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart