AC5394386206 Accela ChemBio Inc.

Methyl 1-Boc-3-ethylpiperidine-3-carboxylate

  • MDL number MFCD21641312
  • Cas Number : 1363165-85-8
  • Molecular Weight : 271.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018684&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018684.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart