AC5394386229 Accela ChemBio Inc.

2-Cyano-3-(dimethylamino)-2-butenamide

  • MDL number MFCD00663777
  • Cas Number : 62321-92-0
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021768&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021768.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart