AC5394386296 Accela ChemBio Inc.

1-Hydroxy-6-(trifluoromethyl)benzotriazole

  • MDL number MFCD00038806
  • Cas Number : 26198-21-0
  • Molecular Weight : 203.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023542&attname=GoodsCode

Safety Information

signalword Danger
pcodes P210-P241-P264-P280-P302+P350-P305+P351+P338-P332+P313-P337+P313-P362
hcodes GHS02,GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY023542.pdf
hstatements H228-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart