AC5394386312 Accela ChemBio Inc.

N-Methylethylenediamine Dihydrochloride

  • MDL number MFCD11846207
  • Cas Number : 64670-85-5
  • Molecular Weight : 147.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023647&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023647.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart