AC5394386319 Accela ChemBio Inc.

1-(2-Pyridyl)ethylamine Dihydrochloride

  • MDL number MFCD09028135
  • Cas Number : 40154-81-2
  • Molecular Weight : 195.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023696&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023696.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart